missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Aisle Marking Tapes
Ultra strong B-726 abrasion-resistant vinyl Aisle Marking Tape makes your facility safer, more visual and more efficient by adding colorful visual indicators to aisles and passageways.
Supplier: Brady™ 58251
Description
- Pressure sensitive rubber based adhesive sticks tightly to virtually any clean, dry surface
- Good chemical, oil and water resistance
- Ideal for marking aisles, passageways and storage locations and identifying equipment and egress paths
- Quick application
Chemical Identifiers
| Acrylic | |
| Vinyl | |
| For pipe markers, arrow tape, warehouse marking, floor marking, warning panels, safety signs and hazardous substance identification | |
| -4°F to 167°F | |
| 0.004 in. | |
| 33 m |
| Tape | |
| Non-sterile | |
| 3 Years | |
| -20°C to +75°C | |
| 36 yd. |
Specifications
| Acrylic | |
| Tape | |
| Marking Tape | |
| For pipe markers, arrow tape, warehouse marking, floor marking, warning panels, safety signs and hazardous substance identification | |
| 1 Roll | |
| -20°C to +75°C | |
| 36 yd. | |
| 3 in. |
| Red | |
| Vinyl | |
| Non-sterile | |
| 3 Years | |
| -4°F to 167°F | |
| 0.004 in. | |
| 33 m | |
| 76.2 mm |