missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Admittance Signs: DANGER - EXPLOSIVES KEEP OUT

Catalog No. 19035950
Change view
Click to view available options
Material:
Fiberglass
Plastic
Temperature Range (English):
-40°F to 175°F
-40°F to 193°F
Thickness:
0.06 in.
0.1 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Temperature Range (English) Thickness
19035950 Plastic -40°F to 175°F 0.06 in.
19035948 Fiberglass -40°F to 193°F 0.1 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19035950 Supplier Brady™ Supplier No. BRD25659

Identify equipment, rooms and materials which contain or are contaminated with hazardous agents

  • Available in three materials - plastic, fiberglass and polyester
  • Black/red on white

Specifications

Material Plastic
Quantity 1 Ea.
Mounting Type Surface Mount
For Use With (Application) Designed for use in industrial, utility, commercial and institutional environments
Height (English) 10 in.
Height (Metric) 25.40 cm
Max. Temperature (English) 175°F
Min. Temperature (English) -40°F
Thickness 0.06 in.
Width (English) 14 in.
Width (Metric) 35.56 cm
Temperature Range (Metric) -40°C to 80°C
Temperature Range (English) -40°F to 175°F
Displayed Text Explosive Keep Out
Product Type Admittance Sign
Color Red on White
Show More Show Less