missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Admittance Signs: DANGER - EXPLOSIVES KEEP OUT

Identify equipment, rooms and materials which contain or are contaminated with hazardous agents

Supplier:  Brady™ 70330

 View more versions of this product

Catalog No. 19035948


Only null left

Description

Description

  • Available in three materials - plastic, fiberglass and polyester
  • Black/red on white

Specifications

Chemical Identifiers

1 Ea.
Designed for use in industrial, utility, commercial and institutional environments
25.40 cm
14 in.
Explosive Keep Out
Surface Mount
10 in.
-40°F
35.56 cm
Admittance Sign

Specifications

Fiberglass
9000 psi
Designed for use in industrial, utility, commercial and institutional environments
25.40 cm
-40°F
14 in.
-40°C to 90°C
Explosive Keep Out
Red on White
1 Ea.
Surface Mount
10 in.
193°F
0.1 in.
35.56 cm
-40°F to 193°F
Admittance Sign
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions