Learn More
Brady™ Accident Prevention Tags - Caution
Accident prevention Caution tags are perfect for marking on-the-spot warnings of dangerous conditions. Available in a variety of different materials and sizes to fit your need.
Supplier:  Brady™ 86430
 View more versions of this product
Catalog No. 19040206
Only null left
Description
Description
  • Rigid economy polyester and cardstock write-on tags meet OSHA Standard 1910.145 for accident prevention. Durable heavy duty write-on polyester (B-851 and B-837) tags with brass grommets exceeds OSHA's 50 lb pull force requirement for tagout devices. Withstands moisture, grease and dirt. Paper cardstock (B-853) tags are recommended for indoor use only.
Specifications
Chemical Identifiers
Black, Yellow
0.37 in.
0.93 cm
Specifications
ASTM D1593-09, ASTM D-638-08, OSHA 1910.145
Polyester
For lockout and tagout identification
3.9 Gardner
14.6cm
0.51 to 0.64 mm
-40°F to 175°F
7.62 cm
0.37 in.
Black, Yellow
Front: Unsafe Do Not Use, Back: Do Not Remove This Tag
2 Years
5.75 in.
10 Nylon Ties
-40°C to 80°C
3 in.
5.75 x 3 in.
0.93 cm
Product Suggestions
Product Suggestions