Learn More
Brady™ 97 Series Indoor/Outdoor Combination Packs
97 Series Combination Packs are B-946 vinyl printed with weather resistant inks and are suited for use in outdoor applications where weatherability is a key consideration.
Supplier:  Brady™ 9713# KIT
 View more versions of this product
Catalog No. 19017789
Only null left
Description
Description
  • Designed for permanent applications in severe indoor or outdoor environments
  • High gloss, durable, flexible vinyl with a permanent cold temperature pressure sensitive acrylic adhesive
  • Good chemical, oil and water resistance
  • Average outdoor durability 8-10 years
Specifications
Chemical Identifiers
RoHS
Vinyl
Permanent Acrylic
180°F
0.004 in.
-40°F to 180°F
General & Industrial ID
Consecutive
Indoor and outdoor use, used for pipemarkers, arrow tape, numbers and letters, vehicle placards, pedestal markers, warning panels and safety signs
-40°F
-40°C to 82°C
Black on White
Specifications
RoHS
Vinyl
Consecutive
Indoor and outdoor use, used for pipemarkers, arrow tape, numbers and letters, vehicle placards, pedestal markers, warning panels and safety signs
180°F
0.004 in.
30.18 mm
-40°F to 180°F
Assorted 0 thru 9
Black on White
General & Industrial ID
10/Card
Permanent Acrylic
1.5 in.
-40°F
1.06 in.
-40°C to 82°C
1.5 x 1.188 in.
0 thru 9 Combo Pack
Product Suggestions
Product Suggestions