Learn More
Bradley™ Halo™ Swing Down Halo Eye/Face Wash
Provides rapid relief to eyes and face injured by chemicals or particulate.
Supplier:  Bradley™ S19274JW
Catalog No. 19-870-022
Only null left
Explore available promotions
Description
Description
  • Compact, Concealed Fixture
  • Chrome-Plated Brass Pipe and Fittings
  • Universal Identification Sign and Inspection Tag Included
  • Exceeds minimum water flow of 3GPM at 30 PSI
  • Halo Sprayhead
  • Durable, Ceramic Valve
Compliance
Complies with ANSI Standard Z358.1-2009
Specifications
Specifications
Swing Down Eye and Face Wash
Product Suggestions
Product Suggestions