Learn More
Bradley™ Deck Mount Side Swing Eyewash
With the Bradley Deck Mount Side Swing Eyewash, you can provide rapid relief to eyes injured by chemicals or particulates.
Supplier: Bradley™ S19274C
Description
- Deck mounting w/locking mechanism
- Chrome-Plated brass pipe and fittings
- Universal identification sign and inspection tag included
- Exceeds minimum water flow of 0.4 GPM at 30 PSI
- Halo sprayhead
Specifications
| ANSI/ISEA Standard Z358.1 | |
| Deck | |
| Side Swing Eyewash |