missing translation for 'onlineSavingsMsg'
Learn More

Br-Mmc (=4-Bromomethyl-7-methoxycoumarin) 98.0+%, TCI America™

Catalog No. p-7118283
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A55511G 1 g
A55515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A55511G Supplier TCI America Supplier No. A55511G
Only null left

Chemical Identifiers

CAS 35231-44-8
Molecular Formula C11H9BrO3
Molecular Weight (g/mol) 269.09
MDL Number MFCD00006869
InChI Key CTENSLORRMFPDH-UHFFFAOYSA-N
Synonym 4-bromomethyl-7-methoxycoumarin, 4-bromomethyl-7-methoxy-2h-chromen-2-one, br-mmc, ccris 7996, 4-bromomethyl-7-methoxychromen-2-one, 2h-1-benzopyran-2-one, 4-bromomethyl-7-methoxy, 4-bromomethyl-7-methoxy-2-oxo-2h-benzopyran, acmc-1agnd, 4-brommethyl-7-methoxy-2h-chromen-2-on, 4-bromomethyl-7-methoxy coumarin
PubChem CID 121894
IUPAC Name 4-(bromomethyl)-7-methoxy-2H-chromen-2-one
SMILES COC1=CC=C2C(CBr)=CC(=O)OC2=C1

Specifications

Melting Point 215°C
Color White-Yellow
Quantity 1 g
UN Number 3261
Formula Weight 269.09
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Br-Mmc (=4-Bromomethyl-7-methoxycoumarin) [for HPLC Labeling]
TSCA No
Recommended Storage Refrigerator