accessibility menu, dialog, popup

UserName

CAS RN 35231-44-8

IUPAC Name: 4-(bromomethyl)-7-methoxy-2H-chromen-2-one
Synonyms: 4-(bromomethyl)-7-methoxychromen-2-one
Molecular Weight (g/mol): 269.09
Molecular Formula: C11H9BrO3
InChi Key: CTENSLORRMFPDH-UHFFFAOYSA-N
SMILES: COC1=CC=C2C(CBr)=CC(=O)OC2=C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)