missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Bovine Insulin ELISA Kit
Description
Bovine Insulin quantitates bovine Insulin in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant bovine Insulin.
Insulin is a peptide hormone secreted by beta cells of the pancreatic islets. It regulates carbohydrate, protein and lipid metabolism by enhancing membrane transport of glucose, amino acids, and certain ions. It also promotes glycogen storage, formation of triglycerides and synthesis of proteins and nucleic acids. Insulin is 51-amino acid polypeptide product produced from a precursor peptide, proinsulin. Proinsulin is post-translationally cleaved into two chains (peptide A and peptide B) that are covalently linked via two disulfide bonds and one molecule of C-peptide. Insulin deficiency results in diabetes mellitus, one of the leading causes of morbidity and mortality in the general population. Insulin is also present in tumors of b-cell origin such as insulinoma.
Specifications
Specifications
| Accession Number | P01308, P01315, P01317, P01322, P01323, P01325, P01326 |
| Assay Range | 4.5-300 μIU/mL |
| Assay Sensitivity | 4.5 μIU/mL |
| Conjugate | HRP |
| Immunoassay Kit Format | Sandwich ELISA Kit |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| Target Species | Human, Mouse, Rat, Bovine, Pig |
| For Use With (Equipment) | Absorbance Microplate Reader |
| Gene Alias | AA986540; IDDM; IDDM1; IDDM2; ILPR; INS; Ins1; Ins-1; Ins2; Ins-2; Ins2-rs1; InsII; Insulin; insulin 1; insulin 2; Insulin A chain; Insulin B chain; insulin I; insulin II; insulin-1; Insulin-1 A chain; Insulin-1 B chain; Insulin-2; Insulin-2 A chain; Insulin-2 B chain; IRDN; Mody; MODY10; Mody4; preproinsulin; proinsulinAA986540; IDDM; IDDM1; IDDM2; ILPR; INS; Ins1; Ins-1; Ins2; Ins-2; Ins2-rs1; InsII; Insulin; insulin 1; insulin 2; Insulin A chain; Insulin B chain; insulin I; insulin II; insulin-1; Insulin-1 A chain; Insulin-1 B chain; Insulin-2; Insulin-2 A chain; Insulin-2 B chain; IRDN; Mody; MODY10; Mody4; preproinsulin; proinsulin; |
| Show More |