Learn More
Invitrogen™ Bovine IL-6 Uncoated ELISA Kit
Description
The Bovine IL-6 Uncoated ELISA Kit contains pre-matched antibody pairs and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of bovine IL-6. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
Specifications
Specifications
| Accession Number | P26892 |
| Assay Range | 78.1 to 5000 pg/mL |
| Assay Sensitivity | <78.1 pg/mL |
| Description | Bovine IL-6 ELISA Reagent Kit |
| Conjugate | HRP |
| Immunoassay Kit Format | Colorimetric ELISA (TMB substrate) with Plates |
| Product Type | ELISA |
| Sample Type | Serum, Supernatant |
| Specificity | No significant cross-reaction with recombinant bovine IL-1ß, IL-2, IL-4, IL-8, IFNg, or TNFa |
| Sufficient For | 5 x 96 microplate wells |
| Show More |