missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Bovine IL-4 ELISA Kit
ELISA
$942.00 - $4,420.00
Specifications
| Accession Number | B cell growth factor 1; B |
|---|---|
| Assay Range | 8 to 2,000 pg/mL |
| Assay Sensitivity | 8 pg/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
EB12RBX5
|
Invitrogen™
EB12RBX5 |
5 x 96 Tests |
N/A
|
|
|||||
EB12RB
|
Invitrogen™
EB12RB |
96 Tests |
N/A
|
|
|||||
Description
Bovine IL-4 quantitates bovine IL-4 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant bovine IL-4.
Interleukin-4 (IL-4) is a pleiotropic, immune-modulatory cytokine produced by activated T cells, and serves as a ligand for interleukin 4 receptor. IL-4 has been found to have the broadest range of actions. The interleukin 4 receptor also binds to IL13, which may contribute to many overlapping functions of this cytokine and IL13. STAT6, a signal transducer and activator of transcription, has been shown to play an important role in mediating the immune regulatory signal of IL-4. The IL-4 gene, IL-3, IL-5, IL-13, and CSF2 form a cytokine gene cluster on chromosome 5q. In particular, IL-4 is located in close proximity to IL13 on the chromosome. Further, the IL-4 gene, IL13 and IL5 are found to be coordinated by several long-range regulatory elements over a 120 kilobase range on the chromosome. IL-4 delta 2 is a known splice variant of IL-4.Specifications
| B cell growth factor 1; B | |
| 8 pg/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| Il4 | |
| <10 | |
| HRP | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| IL-4 | |
| 4 hrs 45 mins |
| 8 to 2,000 pg/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 280824 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |