missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Accession Number | CVID11; C x 2; il 21; Il21; IL-21; ILN; Interleukin; interleukin 21; interleukin 21-like; Interleukin21; interleukin-21; interleukin-21 isoform; LOC100344172; porcine interleukin-21; Za11 |
|---|---|
| Assay Range | 0.41 to 100 ng/mL |
| Assay Sensitivity | 0.41 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
EB11RBX5
|
Invitrogen™
EB11RBX5 |
5 x 96 Tests |
Each for $4,420.00
|
|
|||||
EB11RB
|
Invitrogen™
EB11RB |
96 Tests |
Each for $938.00
|
|
|||||
Description
Bovine IL-21 quantitates bovine IL-21 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant bovine IL-21.
IL-21 is a 17 kDa immunomodulatory cytokine produced mainly by Natural Killer Cells (NKT), T helper (Th) 17 and T follicular helper (TFH) cells. In TFH cells, IL-21 expression leads to autocrine signaling through the IL-21 receptor (IL-21R) and STAT3, which leads to additional transcriptional activation by Bcl6. As with IFN gamma for Th1 and IL-4 for Th2 cells, IL-21 is critical for TFH cell development and effector function. IL-21 plays a role in T cell-dependent B cell differentiation into plasma cells and memory cells, stimulation of IgG production and induction of apoptotic signaling in naive B cells. In Th17 cells, IL-21 expression and autocrine feedback through STAT3, IRF4 and ROR gamma t lead to upregulation of the IL-23R, thereby preparing Th17 cells for maturation and maintenance by the inflammatory cytokine IL-23. While upregulating IRF4 and ROR gamma t, IL-21 also mediates the downregulation of Foxp3. IL-21 deficient mice are protected from developing colitis upon chemical treatment by their inability to upregulate Th17-associated molecules. In comparison, elevated levels of IL-21 are present in chemically-induced colitis mouse models.Specifications
| CVID11; C x 2; il 21; Il21; IL-21; ILN; Interleukin; interleukin 21; interleukin 21-like; Interleukin21; interleukin-21; interleukin-21 isoform; LOC100344172; porcine interleukin-21; Za11 | |
| 0.41 ng/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| IL21 | |
| <10 | |
| HRP | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| IL-21 | |
| 4 hrs 45 mins |
| 0.41 to 100 ng/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 378475 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |