missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Bovine IL-17A ELISA Kit
ELISA
Supplier: Invitrogen™ EB10RB
Description
Bovine IL-17A quantitates bovine IL-17A in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant bovine IL-17A.
Interleukin-17A (IL-17A, CTLA-8) is a CD4+ T cell-derived cytokine that promotes inflammatory responses in cell lines and is elevated in rheumatoid arthritis, asthma, multiple sclerosis, psoriasis, and transplant rejection. IL-17A is a 32 kDa long, disulfide-linked homodimer consisting of 136 amino acids that is a member of a six-species family of proteins (IL-17A-17F) and signals through the IL-17 receptor (IL-17R/CDw217). High levels of IL-17A homodimer are produced by activated peripheral blood CD4+ T-cells, and IL-17A also enhances expression of the intracellular adhesion molecule-1 (ICAM-1) in human fibroblasts. In particular, human IL-17A also stimulates epithelial, endothelial, or fibroblastic cells to secrete IL-6, IL-8, G-CSF, and PGE2. In the presence of human IL-17A, fibroblasts can sustain the proliferation of CD34+ hematopoietic progenitors and induce maturation into neutrophils. Mouse, rat, and human IL-17A can induce IL-6 secretion in mouse stromal cells, indicating that all homologs can recognize the mouse receptor. IL-17A regulates the activities of NF-kappa B and mitogen-activated protein kinases, stimulates the expression of IL-6 and cyclooxygenase-2 (PTGS2/COX-2), and enhances the production of nitric oxide (NO).Chemical Identifiers
| CTLA8; Ctla-8; cytokine; cytotoxic T-lymphocyte-associated antigen 8; cytotoxic T-lymphocyte-associated protein 8; il 17; Il17; IL-17; Il17a; IL-17 A; ILN; Interleukin; interleukin 17; Interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8); interleukin 17 precursor; interleukin 17 A; interleukin 17 A-like; interleukin-17; Interleukin17A; interleukin-17 A; LOC100339322; R-IL-17-A | |
| 2 pg/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| IL17A | |
| <10 | |
| HRP | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| IL-17 A | |
| 4 hrs 45 mins |
| 2.05 to 500 pg/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 282863 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |
Specifications
| CTLA8; Ctla-8; cytokine; cytotoxic T-lymphocyte-associated antigen 8; cytotoxic T-lymphocyte-associated protein 8; il 17; Il17; IL-17; Il17a; IL-17 A; ILN; Interleukin; interleukin 17; Interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8); interleukin 17 precursor; interleukin 17 A; interleukin 17 A-like; interleukin-17; Interleukin17A; interleukin-17 A; LOC100339322; R-IL-17-A | |
| 2 pg/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| IL17A | |
| <10 | |
| HRP | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |
| 2.05 to 500 pg/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 282863 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| 96 Tests | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| IL-17 A | |
| 4 hrs 45 mins |