missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Bovine IFN gamma Uncoated ELISA Kit

Catalog No. ENESS0026B
Encompass
Change view
Click to view available options
Quantity:
5 x 96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ENESS0026B 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ENESS0026B Supplier Invitrogen™ Supplier No. ESS0026B

ELISA

The Bovine IFN-gamma Uncoated ELISA Kit contains pre-matched antibody pairs and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of bovine IFN gamma. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.

IFN gamma (Interferon gamma, Type II interferon) is a macrophage activation factor, and immune interferon that is produced primarily by T-lymphocytes and natural killer cells in response to antigens, mitogens, Staphylococcus enterotoxin B, phytohemaglutanin and other cytokines. IFN gamma is a dimeric protein consisting of two 146 amino acid subunits. IFN gamma is a glycoprotein that exists, functionally, as a homodimer of approximately 45 kDa. On SDS-PAGE, IFN gamma appears as a combination of 25, 20 and minor 15.5 kDa bands as a result of differential glycosylation. The biological activity of the IFN gamma homodimer is highly species specific. Human IFN gamma does not show cross-reactivity with mouse. IFN gamma function includes the following: antiviral activity, tumor antiproliferative activity, induction of class I and II MHC, macrophage activation, and enhanced immunoglobulin secretion by B lymphocytes. IFN gamma is involved in cytokine regulation and also acts synergistically with other cytokines. Activation of IFN gamma takes place through binding of IFN gamma receptor I and II, and activating the JAK-STAT pathway. IFN gamma does not show any homology with IFN alpha or IFN beta but human IFN gamma shows about 40% sequence homology with mouse IFN gamma. IFN gamma is upregulated by IL2, FGF basic, EGF and downregulated by vitamin D3 or DMN. IFN gamma gene mutations are associated with aplastic anemia.

Specifications

Accession Number P07353
Assay Range 31.3 to 2000 pg/mL
Assay Sensitivity <31.3 pg/mL
Description Bovine IFN gamma ELISA Reagent Kit
Conjugate HRP
Immunoassay Kit Format Colorimetric ELISA (TMB substrate) with Plates
Product Type ELISA
Sample Type Serum, Supernatant
Specificity No significant cross-reaction with recombinant bovine IL-1ß, IL-2, IL-4, IL-6, IL-8, or TNFa, or with recombinant human, mouse or rat IFNg
Sufficient For 5 x 96 microplate wells
Target Species Bovine
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 281237
Gene Symbol IFNG
Kit Contents Coating Antibody, Lyophilized Recombinant Standard, Detection Antibody, Streptavidin-HRP, Substrate Solution, Ready to use Stop Solution, 0.16M Sulfuric Acid, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards
Label or Dye HRP
Measurement Absorbance at 450 nm minus absorbance (at 550 nm)
Precision Not measured
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Serum 100 μL, Supernatant 100 μL
Storage Requirements 2°C to 8°C
Target IFN gamma
Test Time 24 hrs
Total Assay Time 2 hrs 50 mins
Show More Show Less