missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Accession Number | alpha-2 A interferon; Gm12597; ifa; Ifa1; Ifa2; IFL; IFN; IFN alpha; IFN a; IFN1@; IFNA; IFNA@; IFNA1; IFNA13; Ifna15; IFNA2; IFNA2A; IFNA2B; IFNA2C; IFNA3; IFNA6; Ifnaa; IFNAalphaIFN-alpha-1/13; IFNalpha; IFN-alpha; IFN-alpha 1 b; IFN-alpha1; IFN-alpha-1; IFN-alpha-1/13; IFN-alpha-2; IFN-alpha-6; IFN-alphaA; IFN-alphaD; IFN-gamma; IFNa; INFA2; interferon; interferon alpha; interferon alpha 1; interferon alpha 15; interferon alpha 1 b; interferon alpha 2; interferon alpha 2 A; interferon alpha 2 b; interferon alpha A; interferon alpha family gene 1; interferon alpha family, gene 2; interferon alpha protein; interferon alpha-1; interferon alpha-1 precursor; Interferon alpha-1/13; interferon alpha-2; Interferon alpha-A; Interferon alpha-D; interferon type A1/A2; interferon type A3; interferon, alpha 1; interferon, alpha 2; interferon-alpha; interferon-alpha 1; interferon-alpha1; interferon-alpha-6; leIF A; LeIF D; MGC138207; MGC138505; MGC138507; MuIFN-alpha-A; novel interferon alpha (Ifna) family member; OTTMUSG00000007655 |
|---|---|
| Assay Range | 0.12 to 30 ng/mL |
| Assay Sensitivity | 0.12 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
EB5RBX5
|
Invitrogen™
EB5RBX5 |
5 x 96 Tests |
Each for $4,420.00
|
|
|||||
EB5RB
|
Invitrogen™
EB5RB |
96 Tests |
Each for $938.00
|
|
|||||
Description
Bovine IFN-alpha quantitates bovine IFN-alpha in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant bovine IFN-alpha.
IFN alpha (Interferon alpha) is a Type I interferon belonging to a family of structurally related, species-specific proteins exclusive to vertebrates. The interaction of IFN-alpha with its receptor component result in the activation of a number of signaling pathways which are mainly involved in innate immune response against viral infection. In humans, the gene for IFN alpha is located on on chromosome 9. The type I interferons consist of 14 different alpha isoforms (subtypes with slightly different specificities), and single beta, omega, epsilon and kappa isoforms. Homologous molecules are found in many species, including rats and mice (and most mammals), and have been identified in birds, reptiles and fish species. IFN alpha is produced primarily by plasmacytoid dendritic cells, is a potent component of the anti-viral innate immune response and modulates adaptive immunity. Diseases associated with the dysfunction of IFN alpha include Hepatitis C and Hepatitis.Specifications
| alpha-2 A interferon; Gm12597; ifa; Ifa1; Ifa2; IFL; IFN; IFN alpha; IFN a; IFN1@; IFNA; IFNA@; IFNA1; IFNA13; Ifna15; IFNA2; IFNA2A; IFNA2B; IFNA2C; IFNA3; IFNA6; Ifnaa; IFNAalphaIFN-alpha-1/13; IFNalpha; IFN-alpha; IFN-alpha 1 b; IFN-alpha1; IFN-alpha-1; IFN-alpha-1/13; IFN-alpha-2; IFN-alpha-6; IFN-alphaA; IFN-alphaD; IFN-gamma; IFNa; INFA2; interferon; interferon alpha; interferon alpha 1; interferon alpha 15; interferon alpha 1 b; interferon alpha 2; interferon alpha 2 A; interferon alpha 2 b; interferon alpha A; interferon alpha family gene 1; interferon alpha family, gene 2; interferon alpha protein; interferon alpha-1; interferon alpha-1 precursor; Interferon alpha-1/13; interferon alpha-2; Interferon alpha-A; Interferon alpha-D; interferon type A1/A2; interferon type A3; interferon, alpha 1; interferon, alpha 2; interferon-alpha; interferon-alpha 1; interferon-alpha1; interferon-alpha-6; leIF A; LeIF D; MGC138207; MGC138505; MGC138507; MuIFN-alpha-A; novel interferon alpha (Ifna) family member; OTTMUSG00000007655 | |
| 0.12 ng/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| IFNA1 | |
| <10 | |
| HRP | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| IFN alpha | |
| 4 hrs 45 mins |
| 0.12 to 30 ng/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 281843 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |