missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Bovine Factor Xa (DEGR) Native Protein

Catalog No. PIRP43120
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP43120 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIRP43120 Supplier Invitrogen™ Supplier No. RP43120

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Native Protein

RP-43120 contains native bovine Factor Xa protein, with the active site blocked by fluorescent inhibitor Dansyl-EGRck. RP-43120 may be used as a positive control in many immunological assays.

Factor Xa is a vitamin K-dependent glycoprotein that converts prothrombin to thrombin in the presence of factor Va, calcium and phospholipid during blood clotting.

Specifications

Accession Number P00743
For Use With (Application) Neutralization
Formulation 50% water with no preservative
Gene ID (Entrez) 280787
Molecular Weight (g/mol) 45.3 kDa
Name Bovine Factor Xa (DEGR)
pH Range 7.4
Purification Method SDS-PAGE
Quantity 100 μg
Source Bovine
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias Activated factor Xa heavy chain; Coagulation factor X; F10; F10a; Factor X heavy chain; Factor X light chain; factor Xa; FX; FXA; prothrombinase; stuart factor; Stuart-Prower factor; Unknown (protein for IMAGE:8121240); unnamed protein product
Common Name Factor Xa
Gene Symbol F10
Product Type Protein
Conjugate Unconjugated
Species Bovine
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Form Liquid
Protein Subtype Native
Show More Show Less