missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Bovine Factor V Native Protein

Catalog No. PIRP43127
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP43127 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIRP43127 Supplier Invitrogen™ Supplier No. RP43127

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Native Protein

RP-43127 contains native bovine Factor V protein. RP-43127 may be used as a positive control in many immunological assays.

Coagulation factor V (or Proaccelerin) is an essential factor of the blood coagulation cascade. This factor circulates in plasma, and is converted to the active form by the release of the activation peptide by thrombin during coagulation. This generates a heavy chain and a light chain which are held together by calcium ions. The active factor V is a cofactor that participates with activated coagulation factor X to activate prothrombin to thrombin. Defects in the Factor V gene can result various diseases including Factor V deficiency, thrombophilia due to activated protein C resistance, Budd-Chiari syndrome, Ischemic stroke, and recurrent pregnancy loss.

Specifications

Accession Number Q28107
For Use With (Application) Neutralization
Formulation 50% water with 50% glycerol and no preservative
Gene ID (Entrez) 280687
Molecular Weight (g/mol) 330 kDa
Name Bovine Factor V
Purification Method SDS-PAGE
Quantity 100 μg
Source Bovine
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias Ac2-120; Activated protein C cofactor; AI173222; Cf5; Cf-5; coagulation factor V; coagulation factor V (proaccelerin, labile factor); Coagulation factor V heavy chain; coagulation factor V jinjiang A2 domain; Coagulation factor V light chain; F5; Factor 5; factor V; factor V Leiden; Factor5; FVL; PCCF; Proaccelerin, labile factor; RPRGL1; THPH2
Common Name Factor V
Gene Symbol F5
Product Type Protein
Conjugate Unconjugated
Species Bovine
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Form Liquid
Protein Subtype Native
Show More Show Less