missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Bovine Factor IX Native Protein

Catalog No. PIRP43070
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP43070 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIRP43070 Supplier Invitrogen™ Supplier No. RP43070

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Native Protein

RP-43070 contains native bovine Factor IX protein. RP-43070 may be used as a positive control in many immunological assays.

Coagulation factor IX is a single chain polypeptide with a molecular mass of 57 kDa. Normal concentrations of factor IX in human plasma is approximately 3 μg/mL. Factor IX deficiency and lack of coagulation activity is seen in patients suffering from the innate X-chromosome linked disorder, haemophilia B.

Specifications

Accession Number P00741
For Use With (Application) Neutralization
Formulation 50% water with 50% glycerol and no preservative
Gene ID (Entrez) 280688
Molecular Weight (g/mol) 55.4 kDa
Name Bovine Factor IX
Purification Method SDS-PAGE
Quantity 100 μg
Source Bovine
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias AW111646; Cf9; Cf-9; Christmas factor; coagulation factor 9; coagulation factor IX; coagulation factor IX (plasma thromboplastic component, Christmas disease, hemophilia B); Coagulation factor IXa heavy chain; Coagulation factor IXa light chain; EC 3.4.21.22; F9; F9 p22; factor 9; factor IX (Christmas factor) precursor; factor IX F9; Factor9; FIX; FIX F9; HEMB; P19; plasma thromboplastic component; plasma thromboplastin component; PTC; RP6-88D7.1; serine protease; THPH8
Common Name Factor IX
Gene Symbol F9
Product Type Protein
Conjugate Unconjugated
Species Bovine
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Protein Subtype Native
Show More Show Less