missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bovine Anti-Rabbit IgG Antibody (H&L) Antibody (biotin), ProSci
Description
Bovine Anti-Rabbit IgG Antibody specifically detects GSK3alpha in Rabbit Samples. It is validated for ELISA, Western Blot, Immunohistochemistry, Immunocytochemistry.
Specifications
Specifications
| Antigen | Bovine Anti-Rabbit IgG |
| Applications | ELISA, Western Blot, Immunohistochemistry, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Biotin |
| Dilution | 1:1,000-1:5,000 for ELISA, Western Blot, color substrates., , 1:5,000-1:10,000 for Western Blot chemiluminescence. |
| Formulation | PBS Buffer containing 0.02% sodium azide. |
| Host Species | Bovine |
| Quantity | 0.5 mg |
| Regulatory Status | RUO |
| Primary or Secondary | Secondary |
| Show More |