missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Bovine anti-Chicken IgY Secondary Antibody, Agarose

Catalog No. SA19588
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SA19588 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SA19588 Supplier Invitrogen™ Supplier No. SA19588
  • ONLINE EXCLUSIVE PRICE
    $1,042.38
    $606.00 / Each

    Save $436.38

    42.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Bovine Polyclonal Secondary Antibody

SA1-9588 contains 1 mL of antibody linked to agarose beads (50%) in PBS with 0.02% sodium azide. This antibody reacts with the heavy and light chains of the immunoglobulin molecule. SA1-9588 immunogen is immunoglobulin isolated from chicken serum. To obtain optimal immunoprecipitation, incubate the beads with samples containing IgY antibodies-protein complex for at least 10 minutes. Binding capacity of the beads is approximately 450 μg to 500 μg IgY per mL solid beads.

Anti-Chicken secondary antibodies are affinity-purified antibodies with well-characterized specificity for the chicken immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Specifications

Antigen Chicken IgY
Applications Immunoprecipitation
Classification Polyclonal
Conjugate Agarose
Formulation PBS with 0.02% sodium azide
Host Species Bovine
Immunogen Immunoglobulin isolated from chicken serum.
Purification Method Protein A/G
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Chicken
Content And Storage 4°C, do not freeze
Product Type Secondary Antibody
Form Slurry
Isotype IgG
Show More Show Less