missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Accession Number | acidic eye-derived fibroblast growth factor; acidic eye-derived growth factor II; Acidic fibroblast growth factor; aFGF; beta-endothelial cell growth factor; Dffrx; ECGF; ECGFA; ECGFB; ECGF-beta; EDGF; EDGF II; Endothelial cell growth factor; Endothelial cell growth factor alpha; Endothelial cell growth factor beta; endothelial cell growth factor beta and alpha chains; endothelial cell growth factor, alpha; endothelial cell growth factor, beta; endothelial growth factor; Fam; FGF; FGF1; Fgf-1; FGF1 protein; Fgfa; FGF-alpha; Fibroblast growth factor; fibroblast growth factor 1; fibroblast growth factor 1 (acidic); Fibroblast growth factor 1 (heparin binding); fibroblast growth factor, acidic; GLIO703; HBGF1; HBGF-1; heparin-binding growth factor; heparin-binding growth factor 1; H-FGF-a; M-FGF-a; Prostatropin; unnamed protein product |
|---|---|
| Assay Range | 0.205 to 50 ng/mL |
| Assay Sensitivity | 0.2 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
EB1RBX5
|
Invitrogen™
EB1RBX5 |
5 x 96 Tests |
Each for $4,420.00
|
|
|||||
EB1RB
|
Invitrogen™
EB1RB |
96 Tests |
Each for $938.00
|
|
|||||
Description
Bovine aFGF/FGF-1 quantitates bovine aFGF/FGF-1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant bovine aFGF/FGF-1.
FGF1 is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. This protein functions as a modifier of endothelial cell migration and proliferation, as well as an angiogenic factor. It acts as a mitogen for a variety of mesoderm- and neuroectoderm-derived cells in vitro, thus is thought to be involved in organogenesis.Specifications
| acidic eye-derived fibroblast growth factor; acidic eye-derived growth factor II; Acidic fibroblast growth factor; aFGF; beta-endothelial cell growth factor; Dffrx; ECGF; ECGFA; ECGFB; ECGF-beta; EDGF; EDGF II; Endothelial cell growth factor; Endothelial cell growth factor alpha; Endothelial cell growth factor beta; endothelial cell growth factor beta and alpha chains; endothelial cell growth factor, alpha; endothelial cell growth factor, beta; endothelial growth factor; Fam; FGF; FGF1; Fgf-1; FGF1 protein; Fgfa; FGF-alpha; Fibroblast growth factor; fibroblast growth factor 1; fibroblast growth factor 1 (acidic); Fibroblast growth factor 1 (heparin binding); fibroblast growth factor, acidic; GLIO703; HBGF1; HBGF-1; heparin-binding growth factor; heparin-binding growth factor 1; H-FGF-a; M-FGF-a; Prostatropin; unnamed protein product | |
| 0.2 ng/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| FGF1 | |
| <10 | |
| HRP | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| FGF1 | |
| 4 hrs 45 mins |
| 0.205 to 50 ng/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 281160 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |