missing translation for 'onlineSavingsMsg'
Learn More

BD Bottled Media TAT Broth

For sterility and non-sterile product testing

$420.95 - $420.95

Specifications

Form Liquid
Manufacturing Origin United States
For Use With (Application) Sterility and non-sterile product testing
Packaging Type Bottle
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
DF9072-73
View Documents
BD
290721
90 mL
Pack of 10 Each for $420.95
Only null left
 
Description

Description

  • For cultivating microorganisms from highly-viscous or gelatinous materials
  • For testing of cosmetics

Specifications

Specifications

Liquid
United States
Sterility and non-sterile product testing
Bottle
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'