Learn More
BD Bottled Media TAT Broth
For sterility and non-sterile product testing
Supplier: BD 290721
Description
- For cultivating microorganisms from highly-viscous or gelatinous materials
- For testing of cosmetics
Specifications
| Liquid | |
| 90 mL | |
| Sterility and non-sterile product testing | |
| 90 mL |
| United States | |
| Pancreatic digest of Casein 20g/L, Soy Lecithin 5g/L, Polysorbate 20 40mL/L | |
| Bottle |