missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Bottle Top Dispensers (BTD) Outer Housing
Outer housing for BTD and dispensing reagents
Specifications
| For Use With (Application) | Dispensing Organic and Inorganic Liquids |
|---|---|
| Includes | Four adapters |
| Catalog Number | Mfr. No. | Volume (Metric) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Volume (Metric) | Price | Quantity & Availability | |||||
|
10320147
|
Corning™
C6846 |
2.5 mL |
N/A
|
|
|||||
|
10320148
|
Corning™
6847 |
5 mL |
N/A
|
|
|||||
|
10320149
|
Corning™
C6848 |
25 mL |
N/A
|
|
|||||
|
10320150
|
Corning™
6849 |
50 mL |
N/A
|
|
|||||
|
10320151
|
Corning™
C6850 |
100 mL |
N/A
|
|
|||||
Description
- Specialized airpurging and closed circuit system ensures no loss of reagent during dispensing, as well as when the device is not in use
- Comes with four adapters with different diameters for universal use with multiple bottle sizes
- Telescopic suction tube can be adjusted smoothly to different bottle heights
- Calibration tool allows easy in-lab calibration with built-in recalibration mechanism to ensure the highest reproducibility during the dispenser’s long life cycle
- Every dispenser is individually tested and delivered with a certificate of performance
- For dispensing organic and inorganic liquids
Specifications
| Dispensing Organic and Inorganic Liquids | |
| Four adapters |