Learn More
Corning™ Bottle Top Dispensers (BTD) Outer Housing
Outer housing for BTD and dispensing reagents
Supplier: Corning™ 6849
Description
- Specialized airpurging and closed circuit system ensures no loss of reagent during dispensing, as well as when the device is not in use
- Comes with four adapters with different diameters for universal use with multiple bottle sizes
- Telescopic suction tube can be adjusted smoothly to different bottle heights
- Calibration tool allows easy in-lab calibration with built-in recalibration mechanism to ensure the highest reproducibility during the dispenser’s long life cycle
- Every dispenser is individually tested and delivered with a certificate of performance
- For dispensing organic and inorganic liquids
Chemical Identifiers
| Dispensing Organic and Inorganic Liquids | |
| Four adapters |
Specifications
| 50 mL | |
| Dispensing Organic and Inorganic Liquids | |
| Four adapters |