missing translation for 'onlineSavingsMsg'
Learn More
Learn More
β-Boswellic acid, ≥95% (HPLC), MilliporeSigma™ Supelco™
Description
Featured Industries: Food and Beverages
Specifications
Specifications
| CAS | 631-69-6 |
| Physical Form | Neat |
| Linear Formula | C30H48O3 |
| MDL Number | MFCD04039448 |
| Synonym | (3 alpha,4 beta)-3-Hydroxyurs-12-en-23-oic acid; 3 alpha-Hydroxyurs-12-en-24-oic acid |
| Type | Analytical Standard |
| Quantity | 5 mg |
| Formula Weight | 456.70 |
| Percent Purity | ≥95% (HPLC) |
| Grade | Analytical Standard |