missing translation for 'onlineSavingsMsg'
Learn More

α-Boswellic acid, ≥95% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014239
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014239 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014239 Supplier Merck Emd Millipore Supplier No. 0681310MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Biologically active pentacyclic triterpene acid suitable for analytical research applications.

Featured Industries: Food and Beverages

Specifications

Chemical Name or Material α-Boswellic Acid
CAS 471-66-9
Linear Formula C30H48O3
Technique Gas Chromatography
MDL Number MFCD03788779
Synonym (3 alpha,4 beta)-3-Hydroxyolean-12-en-23-oic acid; 3 alpha-Hydroxyolean-12-en-24-oic acid
Type Analytical Standard
Molecular Weight (g/mol) 456.70
Industry Type Food and Beverage
Quantity 10 mg
Formula Weight 456.70
Percent Purity ≥95% (HPLC)
Grade Analytical Standard
Show More Show Less