missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bosentan Monohydrate 98.0+%, TCI America™
$92.41 - $271.44
Chemical Identifiers
| CAS | 157212-55-0 |
|---|---|
| Molecular Formula | C27H31N5O7S |
| Molecular Weight (g/mol) | 569.633 |
| MDL Number | MFCD09751188 |
| InChI Key | SXTRWVVIEPWAKM-UHFFFAOYSA-N |
| PubChem CID | 185462 |
| ChEBI | CHEBI:31300 |
| IUPAC Name | 4-tert-butyl-N-[6-(2-hydroxyethoxy)-5-(2-methoxyphenoxy)-2-pyrimidin-2-ylpyrimidin-4-yl]benzenesulfonamide;hydrate |
| SMILES | CC(C)(C)C1=CC=C(C=C1)S(=O)(=O)NC2=C(C(=NC(=N2)C3=NC=CC=N3)OCCO)OC4=CC=CC=C4OC.O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B511825MG
|
TCI America
B511825MG |
25 mg |
Each for $92.41
|
|
|||||
|
B5118100MG
|
TCI America
B5118100MG |
100 mg |
Each for $271.44
|
|
|||||
Chemical Identifiers
| 157212-55-0 | |
| 569.633 | |
| SXTRWVVIEPWAKM-UHFFFAOYSA-N | |
| CHEBI:31300 | |
| CC(C)(C)C1=CC=C(C=C1)S(=O)(=O)NC2=C(C(=NC(=N2)C3=NC=CC=N3)OCCO)OC4=CC=CC=C4OC.O |
| C27H31N5O7S | |
| MFCD09751188 | |
| 185462 | |
| 4-tert-butyl-N-[6-(2-hydroxyethoxy)-5-(2-methoxyphenoxy)-2-pyrimidin-2-ylpyrimidin-4-yl]benzenesulfonamide;hydrate |
Specifications
| 157212-55-0 | |
| C27H31N5O7S | |
| 25 mg | |
| SXTRWVVIEPWAKM-UHFFFAOYSA-N | |
| 4-tert-butyl-N-[6-(2-hydroxyethoxy)-5-(2-methoxyphenoxy)-2-pyrimidin-2-ylpyrimidin-4-yl]benzenesulfonamide;hydrate | |
| 185462 | |
| 551.62 | |
| Crystalline Powder |
| White | |
| MFCD09751188 | |
| 4-(tert-Butyl)-N-[6-(2-hydroxyethoxy)-5-(2-methoxyphenoxy)[2,2′C-bipyrimidin]-4-yl]benzenesulfonamide, 4-tert-Butyl-N-[6-(2-hydroxyethoxy)-5-(2-methoxyphenoxy)-2-(pyrimidin-2-yl)pyrimidin-4-yl]benzenesulfonamide | |
| CC(C)(C)C1=CC=C(C=C1)S(=O)(=O)NC2=C(C(=NC(=N2)C3=NC=CC=N3)OCCO)OC4=CC=CC=C4OC.O | |
| 569.633 | |
| CHEBI:31300 | |
| ≥98.0% (HPLC) | |
| Bosentan Monohydrate |
Safety and Handling
TSCA : No