missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bosentan Monohydrate 98.0+%, TCI America™
Supplier: TCI America B5118100MG
Specifications
| Bosentan Monohydrate | |
| White | |
| MFCD09751188 | |
| 4-(tert-Butyl)-N-[6-(2-hydroxyethoxy)-5-(2-methoxyphenoxy)[2,2′-bipyrimidin]-4-yl]benzenesulfonamide, 4-tert-Butyl-N-[6-(2-hydroxyethoxy)-5-(2-methoxyphenoxy)-2-(pyrimidin-2-yl)pyrimidin-4-yl]benzenesulfonamide | |
| CC(C)(C)C1=CC=C(C=C1)S(=O)(=O)NC2=C(C(=NC(=N2)C3=NC=CC=N3)OCCO)OC4=CC=CC=C4OC.O | |
| 569.633 | |
| CHEBI:31300 | |
| ≥98.0% (HPLC) |
| 157212-55-0 | |
| C27H31N5O7S | |
| 100 mg | |
| SXTRWVVIEPWAKM-UHFFFAOYSA-N | |
| 4-tert-butyl-N-[6-(2-hydroxyethoxy)-5-(2-methoxyphenoxy)-2-pyrimidin-2-ylpyrimidin-4-yl]benzenesulfonamide;hydrate | |
| 185462 | |
| 551.62 | |
| Crystalline Powder |
Chemical Identifiers
| 157212-55-0 | |
| 569.633 | |
| SXTRWVVIEPWAKM-UHFFFAOYSA-N | |
| CHEBI:31300 | |
| CC(C)(C)C1=CC=C(C=C1)S(=O)(=O)NC2=C(C(=NC(=N2)C3=NC=CC=N3)OCCO)OC4=CC=CC=C4OC.O |
| C27H31N5O7S | |
| MFCD09751188 | |
| 185462 | |
| 4-tert-butyl-N-[6-(2-hydroxyethoxy)-5-(2-methoxyphenoxy)-2-pyrimidin-2-ylpyrimidin-4-yl]benzenesulfonamide;hydrate |
Safety and Handling
TSCA : No