missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boscalid 98.0+%, TCI America™
$124.89 - $715.79
Chemical Identifiers
| CAS | 188425-85-6 |
|---|---|
| Molecular Formula | C18H12Cl2N2O |
| Molecular Weight (g/mol) | 343.207 |
| MDL Number | MFCD06795150 |
| InChI Key | WYEMLYFITZORAB-UHFFFAOYSA-N |
| Synonym | 2-Chloro-N-(4′C-chlorobiphenyl-2-yl)nicotinamide |
| PubChem CID | 213013 |
| ChEBI | CHEBI:81822 |
| IUPAC Name | 2-chloro-N-[2-(4-chlorophenyl)phenyl]pyridine-3-carboxamide |
| SMILES | C1=CC=C(C(=C1)C2=CC=C(C=C2)Cl)NC(=O)C3=C(N=CC=C3)Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B4038100MG
|
TCI America
B4038100MG |
100 mg |
Each for $124.89
|
|
|||||
|
B40381G
|
TCI America
B40381G |
1 g |
Each for $715.79
|
|
|||||
Chemical Identifiers
| 188425-85-6 | |
| 343.207 | |
| WYEMLYFITZORAB-UHFFFAOYSA-N | |
| 213013 | |
| 2-chloro-N-[2-(4-chlorophenyl)phenyl]pyridine-3-carboxamide |
| C18H12Cl2N2O | |
| MFCD06795150 | |
| 2-Chloro-N-(4′C-chlorobiphenyl-2-yl)nicotinamide | |
| CHEBI:81822 | |
| C1=CC=C(C(=C1)C2=CC=C(C=C2)Cl)NC(=O)C3=C(N=CC=C3)Cl |
Specifications
| 188425-85-6 | |
| White | |
| MFCD06795150 | |
| 3077 | |
| WYEMLYFITZORAB-UHFFFAOYSA-N | |
| 2-chloro-N-[2-(4-chlorophenyl)phenyl]pyridine-3-carboxamide | |
| 213013 | |
| 343.21 | |
| Crystalline Powder |
| 149°C | |
| C18H12Cl2N2O | |
| 100 mg | |
| 2-Chloro-N-(4′C-chlorobiphenyl-2-yl)nicotinamide | |
| C1=CC=C(C(=C1)C2=CC=C(C=C2)Cl)NC(=O)C3=C(N=CC=C3)Cl | |
| 343.207 | |
| CHEBI:81822 | |
| ≥98.0% (HPLC,N) | |
| Boscalid |
Safety and Handling
RTECSNumber : US4587550
TSCA : No