missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSC Go Science Crazy Borosilicate Glass Tubing, 48 in. length
Constructed from borosilicate glass for greater heat tolerance. GSC International Borosilicate Glass Tubing is available in a variety of diameters for different applications.
$41.49
Specifications
| Material | Borosilicate Glass (3.3) |
|---|---|
| Overall Length (English) | 48 in. |
Alert:
Intended for ages 13 and up.
| Catalog Number | Mfr. No. | Diameter (Metric) Outer | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Diameter (Metric) Outer | Price | Quantity & Availability | |||||
|
S13848
|
GSC Go Science Crazy
10MMBT481 |
10 mm |
Each for $41.49
Pack of 5 Each for $207.45
|
|
|||||
Description
- Sold in one pound packs
- Available in a variety of diameters
Specifications
| Borosilicate Glass (3.3) | |
| 48 in. |