Learn More
Fisherbrand™ Borosilicate Cylinders with Pouring Lip
Heavy-wall construction resists breakage
 View more versions of this product
Catalog No. 08-530C
Only null left
Description
Description
Heavy-wall construction resists breakage
  • Ungraduated
  • Fused round base
  • For use with hydrometers
Specifications
Specifications
Round
Ungraduated Cylinder
4.31 in.
41 mm
Borosilicate Glass
Round
150 mm
1.61 in.
125 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Accessories