missing translation for 'onlineSavingsMsg'
Learn More

Boron triiodide, 95%

Catalog No. p-4781818
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA8722003 1 g
AA8722006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA8722003 Supplier Thermo Scientific Chemicals Supplier No. 08722003
Only null left

CAS: 13517-10-7 | BI3 | 391.52 g/mol

Boron triiodide is used as pharmaceutical, chemical and organic intermediate. It is also used as a catalyst and for preparation of boron compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13517-10-7
Molecular Formula BI3
Molecular Weight (g/mol) 391.52
MDL Number MFCD00036286
InChI Key YMEKEHSRPZAOGO-UHFFFAOYSA-N
Synonym boron triiodide, borane, triiodo, triiodoboron, acmc-1bo2g, boron triiodide 1g, inchi=1/bi3/c2-1 3
PubChem CID 83546
IUPAC Name triiodoborane
SMILES IB(I)I

Specifications

Boiling Point 210°C
Melting Point 48°C to 51°C
Color White
Physical Form Crystals
Quantity 1 g
Assay Percent Range >98%
Solubility Information Reacts with water.
Formula Weight 391.52
Odor Pungent
Sensitivity Light and moisture sensitive
Density 3.35
Chemical Name or Material Boron triiodide
Show More Show Less
Hazard Category H302-H314-H335
Hazard Statement Keep container in a well ventilated place. Keep away from combustible material. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation DOT Class: 8, Packaging Group: II
EINECSNumber 236-857-4
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only