missing translation for 'onlineSavingsMsg'
Learn More

Boron trifluoride dimethanol complex, 50-52 wt% BF3

CAS: 2802-68-8 | C2H8BF3O2 | 131.89 g/mol

$36.83 - $234.02

Chemical Identifiers

CAS 2802-68-8
Molecular Formula C2H8BF3O2
Molecular Weight (g/mol) 131.89
MDL Number MFCD00071635
InChI Key MPRMFRXCVMCOMX-UHFFFAOYSA-N
Synonym boron trifluoride-methanol solution, boron trifluoride-methanol complex, boron trifluoride methanol, boron trifluoride-methanol solution in methanol, bf3 methanol, .bf3 in methanol, boron fluoride methanol, bf3 meoh, borontrifluoride methanol, boron trifluoride solution
PubChem CID 11062313
SMILES CO.CO.FB(F)F
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC158900500
SDS View Documents
Thermo Scientific Chemicals
158900500
50 g Glass bottle
Each for $36.83
Only null left
 
AC158902500
SDS View Documents
Thermo Scientific Chemicals
158902500
250 g Glass bottle
Each for $79.70
Only null left
 
AC158900010
SDS View Documents
Thermo Scientific Chemicals
158900010
1 kg Glass bottle
Each for $234.02
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
Specifications

Chemical Identifiers

2802-68-8
131.89
MPRMFRXCVMCOMX-UHFFFAOYSA-N
11062313
C2H8BF3O2
MFCD00071635
boron trifluoride-methanol solution, boron trifluoride-methanol complex, boron trifluoride methanol, boron trifluoride-methanol solution in methanol, bf3 methanol, .bf3 in methanol, boron fluoride methanol, bf3 meoh, borontrifluoride methanol, boron trifluoride solution
CO.CO.FB(F)F

Specifications

2802-68-8,7637-07-2,67-56-1
Colorless to Yellow
59.0°C (4.0 mmHg)
Glass bottle
1.34
MFCD00071635
1.222
Solubility in water: hydrolysis.
CO.CO.FB(F)F
131.89
131.89
Liquid
-20.0°C
1.2220g/mL
68°C
C2H8BF3O2
BF3·2CH3OH
50 g
boron trifluoride-methanol solution, boron trifluoride-methanol complex, boron trifluoride methanol, boron trifluoride-methanol solution in methanol, bf3 methanol, .bf3 in methanol, boron fluoride methanol, bf3 meoh, borontrifluoride methanol, boron trifluoride solution
MPRMFRXCVMCOMX-UHFFFAOYSA-N
methanol;trifluoroborane
11062313
≥50% (BF3)
Boron trifluoride dimethanol complex, 50-52 wt% BF3
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Causes damage to organs through prolonged or repeated exposure.<br/

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 220-543-9

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only