missing translation for 'onlineSavingsMsg'
Learn More

Boron trifluoride diethyl etherate, 98+%

CAS: 109-63-7 | C4H10BF3O | 141.93 g/mol

Supplier:  Thermo Scientific Chemicals A15275AP

 View more versions of this product

Catalog No. AAA15275AP


Only null left
Explore available promotions

Description

Description

Boron trifluoride diethyl etherate is used as a Lewis acid catalyst in Mukaiyama aldol addition, alkylation, acetylation, isomerization, dehydrations and condensation reactions. It is involved in the prepattion of polyethers in polymerization reactions. As a catalyst, it is used in the preparation of cyclopentyl- and cycloheptyl[b]indoles and other diborane. It is also used in sensitive neutron detectors in ionization chambers as well as monitoring radiation levels in earth’s atmosphere.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Boron trifluoride diethyl etherate is used as a Lewis acid catalyst in Mukaiyama aldol addition, alkylation, acetylation, isomerization, dehydrations and condensation reactions. It is involved in the prepattion of polyethers in polymerization reactions. As a catalyst, it is used in the preparation of cyclopentyl- and cycloheptyl[b]indoles and other diborane. It is also used in sensitive neutron detectors in ionization chambers as well as monitoring radiation levels in earth’s atmosphere.

Solubility
Miscible with ether and alcohol.

Notes
Moisture sensitive. Store in cool place. Recommended to use under inet atomosphere. Incompatible with metals, acids, bases, alcohols, alkali metals, oxidizing agents and water. Reacts violently with water.
Specifications

Specifications

Boron trifluoride diethyl etherate
-60°C
125°C to 126°C
Acrid
1.344
MFCD00013194
UN2604
Moisture sensitive
Boron trifluoride etherate; Boron fluoride-ether
KZMGYPLQYOPHEL-UHFFFAOYSA-N
ethoxyethane;trifluoroborane
8000
≥98%
109-63-7
1.13
47°C (116°F)
C4H10BF3O
BF3€O(CH2CH3)2
500 mL
3909607
14,1350
Miscible with ether and alcohol.
FB(F)F.CCOCC
141.93
141.93

Chemical Identifiers

109-63-7
141.93
KZMGYPLQYOPHEL-UHFFFAOYSA-N
8000
FB(F)F.CCOCC
C4H10BF3O
MFCD00013194
Boron trifluoride etherate; Boron fluoride-ether
ethoxyethane;trifluoroborane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H372-H314-H318-H226-H302
Toxic if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Harmful if swallowed.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H226-H314-H332-H372

DOTInformation : Transport Hazard Class: 8; Packing Group: I; Proper Shipping Name: BORON TRIFLUORIDE DIETHYL ETHERATE

EINECSNumber : 203-689-8

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only