missing translation for 'onlineSavingsMsg'
Learn More

Boron Trifluoride, 14% in Methanol, Spectrum™ Chemical

B2644, 373-57-9, CH4BF3O

Supplier:  Spectrum Chemical Mfg Cor B2644250ML

 View more versions of this product

Catalog No. 18601061


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Boron Trifluoride, 14 Percent in Methanol is used as a reagent in organic chemistry most often as a Lewis acid. It is known to be a versatile building block in several other boron compounds. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Specifications

Specifications

373-57-9
250 mL
MFCD00011316
WTEOIRVLGSZEPR-UHFFFAOYSA-N
trifluoroborane
Reagent
100%
BF3
UN3286
FB(F)F
67.81
Amber Glass Bottle

Chemical Identifiers

373-57-9
67.81
WTEOIRVLGSZEPR-UHFFFAOYSA-N
FB(F)F
BF3
MFCD00011316
trifluoroborane
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions