missing translation for 'onlineSavingsMsg'
Learn More

Boron Trichloride (ca. 9% in Dichloromethane, ca. 1.0mol/L), TCI America™

Supplier:  TCI America B3556100ML

Catalog No. B3556100ML

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Boron Trichloride (ca. 9% in Dichloromethane, ca. 1.0mol/L)
Yellow
MFCD00011313
3265
FAQYAMRNWDIXMY-UHFFFAOYSA-N
trichloroborane
25135
Liquid
10294-34-5
BCl3
100 mL
boron trichloride, borane, trichloro, boron chloride, trichloroboron, borontrichloride, chlorure de bore french, unii-k748471rag, hsdb 326, boron trichloride un1741 poison gas
ClB(Cl)Cl
117.16
117.16

Chemical Identifiers

10294-34-5
117.16
FAQYAMRNWDIXMY-UHFFFAOYSA-N
25135
ClB(Cl)Cl
BCl3
MFCD00011313
boron trichloride, borane, trichloro, boron chloride, trichloroboron, borontrichloride, chlorure de bore french, unii-k748471rag, hsdb 326, boron trichloride un1741 poison gas
trichloroborane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (1)-0042

RTECSNumber : ED1925000

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions