missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boron trichloride, 1M solution in methylene chloride, AcroSeal™
CAS: 10294-34-5 | BCl3 | 117.16 g/mol
$198.64 - $755.52
Specifications
| Chemical Name or Material | Boron trichloride |
|---|---|
| Color | Colorless to Brown |
| Name Note | 1M Solution in Methylene Chloride |
| CAS | 10294-34-5,75-09-2 |
| Molecular Formula | BCl3 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC176681000
|
Thermo Scientific Chemicals
176681000 |
100 mL | AcroSeal™ Glass bottle |
Each for $198.64
|
|
||||
|
AC176688000
|
Thermo Scientific Chemicals
176688000 |
800 mL | AcroSeal™ Glass Bottle |
Each for $755.52
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acidSpecifications
| Boron trichloride | |
| 1M Solution in Methylene Chloride | |
| BCl3 | |
| MFCD00011313 | |
| 01,67; 02,34; 03,31; 04,42; 05,50; 06,65; 09,62; 13,43; 15,44 | |
| 15, 1350 | |
| Solubility in water: may decompose. | |
| ClB(Cl)Cl | |
| 117.16 | |
| 117.17 | |
| Liquid |
| Colorless to Brown | |
| 10294-34-5,75-09-2 | |
| BCl3 | |
| 0.9 to 1.1M | |
| 1.3260g/mL | |
| boron trichloride, borane, trichloro, boron chloride, trichloroboron, borontrichloride, chlorure de bore french, unii-k748471rag, hsdb 326, boron trichloride un1741 poison gas | |
| FAQYAMRNWDIXMY-UHFFFAOYSA-N | |
| trichloroborane | |
| 25135 | |
| 1.326 |
RUO – Research Use Only