missing translation for 'onlineSavingsMsg'
Learn More

Boron trichloride, 1M soln. in hexanes, mixed isomers

Catalog No. AAH32833AE
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH32833AE 100 mL
AAH32833AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH32833AE Supplier Thermo Scientific Chemicals Supplier No. H32833AE

CAS: 10294-34-5 | BCl3 | 117.16 g/mol

A mild, selective combination reagent for the cleavage of primary alkyl aryl ethers. Boron trichloride is a selective demethylating agent. For preparing methyl esters of fatty acids and for transesterification of triglycerides. Mainly used as semiconductor silicon doping source or organic synthesis catalyst, also used for making high purity boron or organic boron.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A mild, selective combination reagent for the cleavage of primary alkyl aryl ethers. Boron trichloride is a selective demethylating agent. For preparing methyl esters of fatty acids and for transesterification of triglycerides. Mainly used as semiconductor silicon doping source or organic synthesis catalyst, also used for making high purity boron or organic boron.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture, heat. Unstable. Store at 2-8°C.

Chemical Identifiers

CAS 10294-34-5
Molecular Formula BCl3
Molecular Weight (g/mol) 117.16
MDL Number MFCD00011313
InChI Key FAQYAMRNWDIXMY-UHFFFAOYSA-N
Synonym boron trichloride, borane, trichloro, boron chloride, trichloroboron, borontrichloride, chlorure de bore french, unii-k748471rag, hsdb 326, boron trichloride un1741 poison gas
PubChem CID 25135
IUPAC Name trichloroborane
SMILES ClB(Cl)Cl

Specifications

Quantity 100 mL
Linear Formula BCl3
UN Number UN3286
Solubility Information Reacts with water.
Formula Weight 117.17
Concentration or Composition (by Analyte or Components) 1M soln. in hexanes
Sensitivity Moisture sensitive; May form a precipitate
Density 0.738
Chemical Name or Material Boron trichloride, mixed isomers
Hazard Category H225-H300+H330-H314-H318-H361-H373-X
Hazard Statement GHS H Statement
H225-H300-H331-H304-H361-H373-H314-H336
Highly flammable liquid and vapor.
Fatal if swallowed.
Toxic if inhaled.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
Precautionary Statement P201-P202-P210-P233-P240-P241-P242-P243-P261-P262-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P310-P330-P331-P363-P372-P374-P380-P501c
DOTInformation Hazard Class: 3; Packaging Group: I
EINECSNumber 233-658-4
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only