missing translation for 'onlineSavingsMsg'
Learn More

Boron trichloride, 1M soln. in dichloromethane, stab.

Catalog No. AAH32751AE
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH32751AE 100 mL
AAH32751AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH32751AE Supplier Thermo Scientific Chemicals Supplier No. H32751AE

CAS: 10294-34-5 | BCl3 | 117.16 g/mol

Boron trichloride is used in semiconductor production and in the preparation of other boron compounds. It serves as a reagent in the preparation of various protease inhibitors. It is also used as a catalyst and soldering flux. It is involved in the improvement of aluminum castings by treating with boron trichloride vapors. It is utilized to produce electrical resistors. It is also used as a metal refining agent, especially for aluminum, magnesium, zinc and copper alloys. Further, it is used in the preparation of nanomaterials such as nanofibers and nanotubes. In addition to this, it is used in the field of high energy fuels and rocket propellants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Boron trichloride is used in semiconductor production and in the preparation of other boron compounds. It serves as a reagent in the preparation of various protease inhibitors. It is also used as a catalyst and soldering flux. It is involved in the improvement of aluminum castings by treating with boron trichloride vapors. It is utilized to produce electrical resistors. It is also used as a metal refining agent, especially for aluminum, magnesium, zinc and copper alloys. Further, it is used in the preparation of nanomaterials such as nanofibers and nanotubes. In addition to this, it is used in the field of high energy fuels and rocket propellants.

Solubility
Miscible with dichloromethane, ethanol, carbon tetrachloride, diethyl ether, dimethyl formamide, aromatic solvents, saturated and halogenated hydrocarbon.

Notes
Moisture sensitive. Incompatible with water, oxidizing agents, bases and metals. Store in a cool place.

Chemical Identifiers

CAS 10294-34-5
Molecular Formula BCl3
Molecular Weight (g/mol) 117.16
MDL Number MFCD00011313
InChI Key FAQYAMRNWDIXMY-UHFFFAOYSA-N
Synonym boron trichloride, borane, trichloro, boron chloride, trichloroboron, borontrichloride, chlorure de bore french, unii-k748471rag, hsdb 326, boron trichloride un1741 poison gas
PubChem CID 25135
IUPAC Name trichloroborane
SMILES ClB(Cl)Cl

Specifications

Quantity 100 mL
UN Number UN2927
Solubility Information Miscible with dichloromethane,ethanol,carbon tetrachloride,diethyl ether,dimethyl formamide,aromatic solvents,saturated and halogenated hydrocarbon.
Formula Weight 117.17
Concentration or Composition (by Analyte or Components) 1M soln.
Sensitivity Moisture sensitive; May form a precipitate
Density 1.326
Chemical Name or Material Boron trichloride, stabilized
Hazard Category H301+H331-H314-H335-H336-H350
Hazard Statement GHS H Statement
H301-H331-H314-H351
Toxic if swallowed.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 233-658-4
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only