Learn More
Boron tribromide, 1M soln. in dichloromethane
CAS: 10294-33-4 | BBr3 | 250.52 g/mol
Supplier: Thermo Scientific Chemicals L14880AC
Description
Boron tribromide is used as a reactant for luminescent polystyrene derivatives with sterically protected carbazolylborane moieties, high-quality boron-doped graphene via Wurtz-type reductive coupling reaction and micrometer-sized organic molecule-DNA hybrid structures. It is also used in bromination and cyclization reactions to prepare cyclopenta[1,3]cyclopropa[1,2-b]pyrrolizin-8-one. It acts as a liquid boron source used in p-type deposition processes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBoron tribromide is used as a reactant for luminescent polystyrene derivatives with sterically protected carbazolylborane moieties, high-quality boron-doped graphene via Wurtz-type reductive coupling reaction and micrometer-sized organic molecule-DNA hybrid structures. It is also used in bromination and cyclization reactions to prepare cyclopenta[1,3]cyclopropa[1,2-b]pyrrolizin-8-one. It acts as a liquid boron source used in p-type deposition processes.
Solubility
Miscible with ethanol and carbon tetrachloride.
Notes
Moisture sensitive. Reacts violently with water. Incompatible with alcohols, alkali metals, aluminum, sodium oxides and strong bases.
Specifications
| Boron tribromide | |
| 25 mL | |
| MFCD00011312 | |
| 14,1347 | |
| Miscible with ethanol and carbon tetrachloride. | |
| BrB(Br)Br | |
| 250.52 | |
| 250.52 | |
| 1.434 | |
| 1.467 |
| 10294-33-4 | |
| BBr3 | |
| UN3390 | |
| boron tribromide, borane, tribromo, boron bromide, tribromoboron, hsdb 327, tribromoboran, boron tribrornide, boron tri bromide, boron tri-bromide | |
| ILAHWRKJUDSMFH-UHFFFAOYSA-N | |
| tribromoborane | |
| 25134 | |
| 1M soln. in dichloromethane | |
| Moisture sensitive |
Chemical Identifiers
| 10294-33-4 | |
| 250.52 | |
| ILAHWRKJUDSMFH-UHFFFAOYSA-N | |
| 25134 | |
| BrB(Br)Br |
| BBr3 | |
| MFCD00011312 | |
| boron tribromide, borane, tribromo, boron bromide, tribromoboron, hsdb 327, tribromoboran, boron tribrornide, boron tri bromide, boron tri-bromide | |
| tribromoborane |
Safety and Handling
GHS H Statement
H300-H330-H314-H351
Fatal if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
P201-P202-P260-P264b-P270-P271-P281-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H300+H330-H314-H335-H336-H350-H373
DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC BY INHALATION LIQUID, CORROSIVE, N.O.S.
EINECSNumber : 233-657-9
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only