missing translation for 'onlineSavingsMsg'
Learn More

Boron Standard For ICP, 1000μg/mL in H2O, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC PLB92X

 View more versions of this product

Catalog No. PLB9-2X

Only null left
Explore available promotions

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
B1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Boron Standard
7440-42-8,7732-18-5
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
0.1,99.9
MFCD00134034 MFCD00151272
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
ZOXJGFHDIHLPTG-UHFFFAOYSA-N
Water
10.81
5462311
Assurance
Triple-leached LDPE bottle
Assurance Certificate of Analysis reports up to 30 elements at ppb levels.
0.1,99.9
B
AA, ICP
Certified Reference Material
[B]
boron
500 mL
CHEBI:27560
Matrix    Concentration    Cat. No.
H2O    1000μg/mL    PLB9-2Y, -2T, -2X    10,000    PLB9-3Y, -3X

Chemical Identifiers

7440-42-8
10.81
ZOXJGFHDIHLPTG-UHFFFAOYSA-N
CHEBI:27560
[B]
B
MFCD00134034 MFCD00151272
5462311
boron
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Causes skin irritation, serious eye irritation. May cause cancer. Suspected of damaging fertility or the unborn child.

DOTInformation : Nonhazardous

Recommended Storage : Ambient

SDS
Documents

Documents

Promotions

Promotions

Product intended for laboratory use only