Learn More
Boron Standard For ICP, 1000μg/mL in H2O, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Supplier: SPEX CertiPrep Group LLC PLB92T
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| B | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
| Boron Standard | |
| 7440-42-8,7732-18-5 | |
| Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
| 0.1,99.9 | |
| MFCD00134034 MFCD00151272 | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| ZOXJGFHDIHLPTG-UHFFFAOYSA-N | |
| Water | |
| 10.81 | |
| 5462311 | |
| Assurance |
| Triple-leached LDPE bottle | |
| Assurance Certificate of Analysis reports up to 30 elements at ppb levels. | |
| 0.1,99.9 | |
| B | |
| AA, ICP | |
| Certified Reference Material | |
| [B] | |
| boron | |
| 250 mL | |
| CHEBI:27560 | |
| Matrix Concentration Cat. No. H2O 1000μg/mL PLB9-2Y, -2T, -2X 10,000 PLB9-3Y, -3X |
Chemical Identifiers
| 7440-42-8 | |
| 10.81 | |
| ZOXJGFHDIHLPTG-UHFFFAOYSA-N | |
| CHEBI:27560 | |
| [B] |
| B | |
| MFCD00134034 MFCD00151272 | |
| 5462311 | |
| boron |
Safety and Handling
Causes skin irritation, serious eye irritation. May cause cancer. Suspected of damaging fertility or the unborn child.
DOTInformation : Nonhazardous
Recommended Storage : Ambient
Product intended for laboratory use only