missing translation for 'onlineSavingsMsg'
Learn More

Boron, plasma standard solution, Specpure™ B 1000μg/mL

$135.08 - $135.08

Specifications

Chemical Name or Material Boron
Name Note plasma standard solution, B 1000μg/mL
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Physical Form Liquid
MDL Number MFCD00134034
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA39147AE
View Documents
Thermo Scientific Chemicals
039147AE
100 mL
Each for $135.08
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Boron
23 hPa (17mm Hg) at 20°C
MFCD00134034
plasma standard solution, B 1000μg/mL
Liquid
Videos
SDS
Documents

Documents

RUO – Research Use Only