missing translation for 'onlineSavingsMsg'
Learn More

Boron pieces, crystalline, 1-30mm (0.039-1.2in), 99.4% (metals basis)

CAS: 7440-42-8 | B | 10.81 g/mol

Supplier:  Thermo Scientific Chemicals 04185818

 View more versions of this product

Catalog No. AA4185818


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Boron
7440-42-8
50 g
B
boron, metallic, unii-n9e3x5056q, boron, amorphous, powder, crystalline, boro, boron-elemental, boron, elemental, and compounds, boracium, boranylidyne
[B]
10.81
CHEBI:27560
Odorless
Gray
Crystalline Pieces
99.4% (Metals basis)
MFCD00134034 MFCD00151272
ZOXJGFHDIHLPTG-UHFFFAOYSA-N
boron
5462311
700°C (1292°F)
Dark Gray pieces

Chemical Identifiers

7440-42-8
10.81
ZOXJGFHDIHLPTG-UHFFFAOYSA-N
5462311
boron
B
MFCD00134034 MFCD00151272
boron, metallic, unii-n9e3x5056q, boron, amorphous, powder, crystalline, boro, boron-elemental, boron, elemental, and compounds, boracium, boranylidyne
CHEBI:27560
[B]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H302; Harmful if swallowed.

EINECSNumber : 231-151-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only