missing translation for 'onlineSavingsMsg'
Learn More

Boron pieces, crystalline, 1-30mm (0.039-1.2in), 99.4% (metals basis)

Catalog No. p-4824635
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4185809 10 g
AA4185818 50 g
AA4185830 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4185809 Supplier Thermo Scientific Chemicals Supplier No. 04185809
Only null left

CAS: 7440-42-8 | B | 10.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-42-8
Molecular Formula B
Molecular Weight (g/mol) 10.81
MDL Number MFCD00134034 MFCD00151272
InChI Key ZOXJGFHDIHLPTG-UHFFFAOYSA-N
Synonym boron, metallic, unii-n9e3x5056q, boron, amorphous, powder, crystalline, boro, boron-elemental, boron, elemental, and compounds, boracium, boranylidyne
PubChem CID 5462311
ChEBI CHEBI:27560
IUPAC Name boron
SMILES [B]

Specifications

Color Gray
Physical Form Crystalline Pieces
Quantity 10 g
Assay Percent Range 99.4% (Metals basis)
Ignition Point 700°C (1292°F)
Odor Odorless
Appearance Dark Gray pieces
Chemical Name or Material Boron
Hazard Category H302; Harmful if swallowed.
EINECSNumber 231-151-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only