missing translation for 'onlineSavingsMsg'
Learn More

Boron phosphate

Catalog No. p-4824634
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3256222 100 g
AA3256236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3256222 Supplier Thermo Scientific Chemicals Supplier No. 03256222

CAS: 13308-51-5 | BO4P | 105.78 g/mol

Boron phosphate is used as a catalyst for dehydration as well as in other organic reactions. It is employed as a source of phosphates involved in the exchange reaction to prepare metal phosphates. It finds application in refractories, ceramics and flame-retardants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13308-51-5
Molecular Formula BO4P
Molecular Weight (g/mol) 105.78
MDL Number MFCD00011318
InChI Key RKVCQBPDVHFKCU-UHFFFAOYSA-N
Synonym boron phosphate, boron orthophosphate, boron phosphate b po4, 2,4,5-trioxa-1-phospha-3-borabicyclo 1.1.1 pentane 1-oxide, 2,4,5-trioxa-1??-phospha-3-borabicyclo 1.1.1 pentan-1-one, borophosphoric acid, boron iii phosphate, boron phosphate hydrate, boronphosphate b po4, boron phosphate 100g
PubChem CID 83329
IUPAC Name 2,4,5-trioxa-1$l^{5}-phospha-3-borabicyclo[1.1.1]pentane 1-oxide
SMILES B12OP(=O)(O1)O2

Specifications

Melting Point >1400°C
Color White
Physical Form Powder
Quantity 100 g
Solubility Information Insoluble in water.
Formula Weight 105.79
Odor Odorless
Density 2.81
Chemical Name or Material Boron phosphate
Hazard Category H302-H360FD
Precautionary Statement P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
EINECSNumber 236-337-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only