missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boron phosphate
Description
Boron phosphate is used as a catalyst for dehydration as well as in other organic reactions. It is employed as a source of phosphates involved in the exchange reaction to prepare metal phosphates. It finds application in refractories, ceramics and flame-retardants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | >1400°C |
| Color | White |
| Physical Form | Powder |
| Quantity | 500 g |
| Solubility Information | Insoluble in water. |
| Formula Weight | 105.79 |
| Odor | Odorless |
| Density | 2.81 |
| Chemical Name or Material | Boron phosphate |
Safety and Handling
| Hazard Category | H302-H360FD |
| Precautionary Statement | P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c |
| EINECSNumber | 236-337-7 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only