missing translation for 'onlineSavingsMsg'
Learn More

Boron oxide, Puratronic™, (H{2}O 200ppm), 99.999% (metals basis), Thermo Scientific Chemicals

$1007.99 - $1007.99

Chemical Identifiers

CAS 1303-86-2
Molecular Formula B2O3
Molecular Weight (g/mol) 69.62
MDL Number MFCD00011315
InChI Key JKWMSGQKBLHBQQ-UHFFFAOYSA-N
Synonym boron trioxide, boric oxide, boric anhydride, boric acid anhydride, oxo oxoboranyloxy borane, boronoxide, diboron trioxide, boron sesquioxide, boron oxide, boron oxide 500g
PubChem CID 518682
ChEBI CHEBI:30163
IUPAC Name oxo(oxoboranyloxy)borane
SMILES O=BOB=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA11160QE
View Documents
Thermo Scientific Chemicals
011160QE
1 cyl
Each for $1,007.99
Only null left
 
Description

Description

In the electronics industry as a liquid encapsulent for Czochralski-type growth of III-V semiconductor single crystals (e.g., GaAs, GaP, InP).Boron oxide is used as a flux, an acid catalyst and a chemical intermediate in the production of boron halides, esters, carbide, nitride and metallic borides. It has shown a promising effect on glycerol hydrogenolysis to 1,2-propanediol. It is also an essential material for the preparation of cordierite ceramics from talc. It is actively involved in the production of borosilicate glass. It is an additive used in high-temperature lubricants, heatproof glasswares and dopants in semiconductor materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • In the electronics industry as a liquid encapsulent for Czochralski-type growth of III-V semiconductor single crystals (e.g., GaAs, GaP, InP)
Specifications

Chemical Identifiers

1303-86-2
69.62
JKWMSGQKBLHBQQ-UHFFFAOYSA-N
518682
oxo(oxoboranyloxy)borane
B2O3
MFCD00011315
boron trioxide, boric oxide, boric anhydride, boric acid anhydride, oxo oxoboranyloxy borane, boronoxide, diboron trioxide, boron sesquioxide, boron oxide, boron oxide 500g
CHEBI:30163
O=BOB=O

Specifications

1860°C
1303-86-2
Cylinder
B2O3
14,1337
Slightly soluble in water and methanol.
O=BOB=O
69.62
CHEBI:30163
99.999%
Odorless
2.46 g/mL (±)0.01
(metals basis)
∼450°C
White
1 cyl
MFCD00011315
boron trioxide, boric oxide, boric anhydride, boric acid anhydride, oxo oxoboranyloxy borane, boronoxide, diboron trioxide, boron sesquioxide, boron oxide, boron oxide 500g
JKWMSGQKBLHBQQ-UHFFFAOYSA-N
oxo(oxoboranyloxy)borane
518682
69.62
Puratronic
Hygroscopic
Boron oxide, H2O 200ppm, Cylinder, 5.0cm dia.
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H360
May damage fertility or the unborn child.

P201-P202-P281-P308+P313-P501c

H360FD

EINECSNumber : 215-125-8

RTECSNumber : ED7900000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only